Anti-ILK Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00003611-A01EA 258 EUR
ABNOH00003611-A01
Anti-ILK Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ILK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR