Anti-GH2 Mouse Monoclonal Antibody [clone: 1D10]

Lieferant: Abnova
ABNOH00002689-M02EA 380 EUR
ABNOH00002689-M02
Anti-GH2 Mouse Monoclonal Antibody [clone: 1D10]
Antikörper
Type: Primary
Antigen: GH2
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR