Anti-SLC29A1 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00002030-B01PEA 394 EUR
ABNOH00002030-B01P
Anti-SLC29A1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC29A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human