Anti-DBN1 Mouse Monoclonal Antibody [clone: 2E11]

Lieferant: Abnova
ABNOH00001627-M03EA 394 EUR
ABNOH00001627-M03
Anti-DBN1 Mouse Monoclonal Antibody [clone: 2E11]
Antikörper
Type: Primary
Antigen: DBN1
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR