Anti-CASP9 Mouse Monoclonal Antibody [clone: 3B8-4G2]

Lieferant: Abnova
ABNOH00000842-M01EA 380 EUR
ABNOH00000842-M01
Anti-CASP9 Mouse Monoclonal Antibody [clone: 3B8-4G2]
Antikörper
Type: Primary
Antigen: CASP9
Clonality: Monoclonal
Clone: 3B8-4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR