Anti-ATP2A1 Mouse Monoclonal Antibody [clone: 5H8]

Lieferant: Abnova
ABNOH00000487-M04EA 380 EUR
ABNOH00000487-M04
Anti-ATP2A1 Mouse Monoclonal Antibody [clone: 5H8]
Antikörper
Type: Primary
Antigen: ATP2A1
Clonality: Monoclonal
Clone: 5H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR