Anti-ATOH1 Mouse Monoclonal Antibody [clone: 3B12]

Lieferant: Abnova
ABNOH00000474-M06EA 380 EUR
ABNOH00000474-M06
Anti-ATOH1 Mouse Monoclonal Antibody [clone: 3B12]
Antikörper
Type: Primary
Antigen: ATOH1
Clonality: Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR