Anti-ASPH Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00000444-D03EA 380 EUR
ABNOH00000444-D03
Anti-ASPH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ASPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR