Anti-APOF Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00000319-B01PEA 380 EUR
ABNOH00000319-B01P
Anti-APOF Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human APOF protein.

Recommended Dilutions: Western Blot (1 µg/ml)

Type: Primary
Antigen: APOF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR