Anti-AMFR Mouse Monoclonal Antibody [clone: 3D9]

Lieferant: Abnova
ABNOH00000267-M01EA 394 EUR
ABNOH00000267-M01
Anti-AMFR Mouse Monoclonal Antibody [clone: 3D9]
Antikörper
Type: Primary
Antigen: AMFR
Clonality: Monoclonal
Clone: 3D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR