Anti-ALCAM Mouse Monoclonal Antibody [clone: 1H3]

Lieferant: Abnova
ABNOH00000214-M02AEA 380 EUR
ABNOH00000214-M02A
Anti-ALCAM Mouse Monoclonal Antibody [clone: 1H3]
Antikörper
Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR