Anti-MAP3K1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314185EA 552 EUR
ORIGTA314185
Anti-MAP3K1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: MAP3K1 (mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR