Anti-EPB41 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313823EA 552 EUR
ORIGTA313823
Anti-EPB41 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: EPB41 (erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR