Anti-CA3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313538EA 552 EUR
ORIGTA313538
Anti-CA3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CA3 (carbonic anhydrase III, muscle specific)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR