Anti-ARHGDIG Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313475EA 532 EUR
ORIGTA313475
Anti-ARHGDIG Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ARHGDIG (Rho GDP dissociation inhibitor (GDI) gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR