Anti-PLCG1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313084EA 552 EUR
ORIGTA313084
Anti-PLCG1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1: 500-1:1000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: PLCG1 (phospholipase C, gamma 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR