Anti-TNFSF9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312998EA 564 EUR
ORIGTA312998
Anti-TNFSF9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: TNFSF9 (tumor necrosis factor (ligand) superfamily, member 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR