Anti-ZC3H8 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312645EA 552 EUR
ORIGTA312645
Anti-ZC3H8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: ZC3H8 (zinc finger CCCH-type containing 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR