Anti-EGFR Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312536EA 552 EUR
ORIGTA312536
Anti-EGFR Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:300-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR