Anti-PPP1R1B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312426EA 564 EUR
ORIGTA312426
Anti-PPP1R1B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: PPP1R1B (protein phosphatase 1, regulatory (inhibitor) subunit 1B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR