Anti-NKX3-1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312302EA 552 EUR
ORIGTA312302
Anti-NKX3-1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: NKX3-1 (NK3 homeobox 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR