Anti-RBMX Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA311681EA 552 EUR
ORIGTA311681
Anti-RBMX Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: RBMX (RNA binding motif protein, X-linked)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR