Anti-TREML1 Goat Antibody

Lieferant: OriGene
ORIGTA311600EA 536 EUR
ORIGTA311600
Anti-TREML1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: TREML1 (triggering receptor expressed on myeloid cells-like 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR