Anti-SYT2 Goat Antibody

Lieferant: OriGene
ORIGTA311599EA 536 EUR
ORIGTA311599
Anti-SYT2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: SYT2 (synaptotagmin II)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR