Anti-OXER1 Goat Antibody

Lieferant: OriGene
ORIGTA311580EA 536 EUR
ORIGTA311580
Anti-OXER1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: OXER1 (oxoeicosanoid (OXE) receptor 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR