Anti-KLHL6 Goat Antibody

Lieferant: OriGene
ORIGTA311566EA 539 EUR
ORIGTA311566
Anti-KLHL6 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: KLHL6 (kelch-like family member 6)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR