Anti-CROT Goat Antibody

Lieferant: OriGene
ORIGTA311543EA 550 EUR
ORIGTA311543
Anti-CROT Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: CROT (carnitine O-octanoyltransferase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR