Anti-ATG16L1 Goat Antibody

Lieferant: OriGene
ORIGTA311536EA 547 EUR
ORIGTA311536
Anti-ATG16L1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.01-0.1?g/ml

Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR