Anti-FANCF Goat Antibody

Lieferant: OriGene
ORIGTA311512EA 539 EUR
ORIGTA311512
Anti-FANCF Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.1-0.5?g/ml

Type: Primary
Antigen: FANCF (Fanconi anemia, complementation group F)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR