Anti-NLGN4X Goat Antibody

Lieferant: OriGene
ORIGTA311492EA 536 EUR
ORIGTA311492
Anti-NLGN4X Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: NLGN4X (neuroligin 4, X-linked)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR