Anti-TERF2IP Goat Antibody

Lieferant: OriGene
ORIGTA311481EA 536 EUR
ORIGTA311481
Anti-TERF2IP Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: TERF2IP (telomeric repeat binding factor 2, interacting protein)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR