Anti-DIAPH1 Goat Antibody

Lieferant: OriGene
ORIGTA311325EA 536 EUR
ORIGTA311325
Anti-DIAPH1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: DIAPH1 (diaphanous-related formin 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR