Anti-MPV17 Goat Antibody

Lieferant: OriGene
ORIGTA311233EA 536 EUR
ORIGTA311233
Anti-MPV17 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: MPV17 (MpV17 mitochondrial inner membrane protein)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR