Anti-MEST Goat Antibody

Lieferant: OriGene
ORIGTA311229EA 536 EUR
ORIGTA311229
Anti-MEST Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.2-0.5?g/ml

Type: Primary
Antigen: MEST (mesoderm specific transcript)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR