Anti-MBOAT4 Goat Antibody

Lieferant: OriGene
ORIGTA311187EA 536 EUR
ORIGTA311187
Anti-MBOAT4 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: MBOAT4 (membrane bound O-acyltransferase domain containing 4)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR