Anti-TRIM71 Goat Antibody

Lieferant: OriGene
ORIGTA311164EA 536 EUR
ORIGTA311164
Anti-TRIM71 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1?g/ml

Type: Primary
Antigen: TRIM71 (tripartite motif containing 71, E3 ubiquitin protein ligase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR