Anti-LPA Rabbit Monoclonal Antibody [clone: EPR6474]

Lieferant: OriGene
ORIGTA310972EA 826 EUR
ORIGTA310972
Anti-LPA Rabbit Monoclonal Antibody [clone: EPR6474]
Antikörper
Recommended Dilutions: Western Blot: 1:10000-1:50000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: LPA (lipoprotein, Lp(a))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR