Anti-AKR1C3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310144EA 359 EUR
ORIGTA310144
Anti-AKR1C3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:50

Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR