Anti-ADCY9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310098EA 385 EUR
ORIGTA310098
Anti-ADCY9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ADCY9 (adenylate cyclase 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR