Anti-LOXL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310063EA 359 EUR
ORIGTA310063
Anti-LOXL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: LOXL1 (lysyl oxidase-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR