Anti-CCR4 Mouse Monoclonal Antibody

Lieferant: OriGene
ORIGTA310022EA 567 EUR
ORIGTA310022
Anti-CCR4 Mouse Monoclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR