Anti-CHEK2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309988EA 603 EUR
ORIGTA309988
Anti-CHEK2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR