Anti-MYL2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309976EA 603 EUR
ORIGTA309976
Anti-MYL2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MYL2 (myosin, light chain 2, regulatory, cardiac, slow)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR