Anti-GATA1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309953EA 574 EUR
ORIGTA309953
Anti-GATA1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: GATA1 (GATA binding protein 1 (globin transcription factor 1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR