Anti-ABCA1 Mouse Monoclonal Antibody [clone: AB.H10]

Lieferant: OriGene
ORIGTA309942EA 566 EUR
ORIGTA309942
Anti-ABCA1 Mouse Monoclonal Antibody [clone: AB.H10]
Antikörper
Recommended Dilutions: Western Blot: 1:1000;

Type: Primary
Antigen: ABCA1 (ATP-binding cassette, sub-family A (ABC1), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Chicken, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR