Anti-HTT Mouse Monoclonal Antibody [clone: H.F5]

Lieferant: OriGene
ORIGTA309937EA 567 EUR
ORIGTA309937
Anti-HTT Mouse Monoclonal Antibody [clone: H.F5]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: HTT (huntingtin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR