Anti-WHSC1L1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309929EA 637 EUR
ORIGTA309929
Anti-WHSC1L1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: WHSC1L1 (Wolf-Hirschhorn syndrome candidate 1-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR