Anti-TAF15 Mouse Monoclonal Antibody [clone: 4D71]
Lieferant: OriGene
ORIGTA309895EA 636 EUR
ORIGTA309895
Anti-TAF15 Mouse Monoclonal Antibody [clone: 4D71]
Antikörper
Recommended Dilutions:
Type: Primary
Antigen: TAF15 (TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human