Anti-TAC3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309892EA 723 EUR
ORIGTA309892
Anti-TAC3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: TAC3 (tachykinin 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR