Anti-SPDYA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309875EA 668 EUR
ORIGTA309875
Anti-SPDYA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SPDYA (speedy/RINGO cell cycle regulator family member A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR